Skip to Content
Merck
All Photos(1)

Key Documents

GF414-M

Sigma-Aldrich

HumanKine® HGF, Human Recombinant Xeno-Free

Synonym(s):

Hepatocyte Growth Factor, Hepatopoeitin-A

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€6,530.00

€6,530.00


Estimated to ship on13 May 2025Details



Select a Size

Change View
1 MG
€6,530.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.27

€6,530.00


Estimated to ship on13 May 2025Details


Assay

>95% (SDS-PAGE)

Quality Level

manufacturer/tradename

HumanKine®

impurities

<0.1 μg/mg protein endotoxin (1 EU/μg)

UniProt accession no.

General description

Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) and Hepatopoietin A, is a pleiotrophic growth factor which is produced by mesodermally derived cells such as endothelial cells, Kupffer cells/macrophages, and hepatic fat-storing cells. HGF/SF is a potent effector of growth, movement, differentiation and invasiveness of epithelial and cancer cells. It can induce epithelial-mesenchymal conversion of epithelial cells in culture, support epithelial branching duct formation in the developing lung and elicit a potent organotrophic function for regeneration of liver, kidney and lung through epithelial-stromal interactions.
HumanKine growth factors are expressed in a proprietary human 293 protein expression system and are manufactured without the use of any animal products and are considered Xeno-Free.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Bioactivity assay: Determined by the dose-dependent stimulation of the proliferation of the monkey epithelial cell line 4MBr-5. ED50 is typically 10-50 ng/ml.

Physical form

Product is presented in PBS and filtered through a 0.2 micron filter before lyophilization.

Preparation Note

HumanKine HGF is manufactured using all non-animal reagents and is considered Xeno-Free

Storage and Stability

Store at -20°C for up to 4 months from date of receipt. Further dilutions should be made into appropriate buffer or medium to which protein has been added.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service