Skip to Content
Merck
All Photos(1)

Key Documents

GF309

Sigma-Aldrich

NT-4 Protein, Human Recombinant Animal Free

The Neurotrophin-4 protein (or NT-4 protein), also known as NT-5 protein, is a member of the NGF family of neuronal & epithelial growth factors.

Synonym(s):

NT4, Neurotrophin 4, Neurotrophin 4/5, NT4/5

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€392.00

€392.00


Estimated to ship on10 April 2025Details



Select a Size

Change View
10 μG
€392.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

€392.00


Estimated to ship on10 April 2025Details


Quality Level

Assay

>98% (SDS-PAGE and HPLC)

impurities

<0.1 μg/mg protein endotoxin (<1EU/μg)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Neurotrophin4 (NT4), also known as NT5, is a member of the NGF family of neuronal and epithelial growth factors. NT-4 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-3. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-4 is expressed in the prostate, thymus, placenta and skeletal muscle. NT-4 can signal through the LNGFR and trkB receptors and promotes the survival of peripheral sensory sympathetic neurons. Recombinant human NT-4 is a noncovalently linked homodimer, of two 14.0 kDa polypeptide monomers (260 total amino acid residues).

Specificity

Cross Reactivty
Mouse, Rat

Quality

Bioactivity assay: The ED50 as determined by the dose-dependent induction of choline acetyl transferase activity in rat basal forebrain primary septal cell cultures was found in the range of 20-50 ng/ml.

Physical form

Recombinant animal free human NT-4 is manufactured using all non-animal reagents

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service