Skip to Content
Merck
All Photos(1)

Key Documents

FXWXM20K0006

Millipore

Mobius® Chrom 20 Flexware® Kit

25mm Post + Pre

Synonym(s):

Mobius® Chrom 20 Flexware® Kit (25mm Post + Pre)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€19,775.00

€19,775.00


Estimated to ship on01 September 2025Details



Select a Size

Change View
1 EACH
€19,775.00

About This Item

UNSPSC Code:
41116107

€19,775.00


Estimated to ship on01 September 2025Details


Quality Level

product line

Mobius®

packaging

pkg of 1 packs

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Legal Information

FLEXWARE is a registered trademark of Merck KGaA, Darmstadt, Germany
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service