Skip to Content
Merck
All Photos(1)

Documents

FCMAB243P

Sigma-Aldrich

Anti-mouse IFN gamma -PE Antibody, clone XMG1.2

clone XMG1.2, from rat, PE

Synonym(s):

IFN-gamma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

XMG1.2, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Ifng(15978)

General description

Interferon gamma is produced by lymphocytes activated by specific antigens or mitogens. IFN-gamma, in addition to having antiviral activity, plays an important role in several immunoregulatory functions. It is a potent activator of macrophages, it has antiproliferative effects on transformed cells, and it can potentiate the antiviral and antitumor effects of the type I interferons.

Specificity

Antibody recognizes Interferon gamma .

Immunogen

Recombinant protein corresponding to E.coli -expressed, recombinant mouse IFN-γ.

Application

Detect mouse IFN gamma using this Anti-mouse IFN gamma -PE Antibody, clone XMG1.2 validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunological Signaling

Quality

Evaluated by flow cytometry using Mick-1 cells

Target description

18 kDa Calculated

Physical form

Affinity purified
Purified mouse monoclonal IgG1κ conjugated to PE in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Mick-1 cells

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service