Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB201F

Sigma-Aldrich

Milli-Mark® Anti-CD57-FITC Antibody, clone TB01

clone TB01, Milli-Mark®, from mouse

Synonym(s):

Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1, Beta-1,3-glucuronyltransferase 1, Glucuronosyltransferase P, GlcAT-P, UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€278.00

€278.00


Estimated to ship on25 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 TESTS
€278.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€278.00


Estimated to ship on25 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

TB01, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... B3GAT1(27087)

General description

The antibody recognizes the CD57 carbohydrate epitope present on several surface molecules. In the nervous system this epitope is particularly prominent on myelin-associated glycoprotein (MAG) (Funaro, 1997). The eptope for TB01 is neuraminidase- and bromelaine-sensitive, but trypsin-resistant (Funaro, 1997).
Anti-CD57, TBO1, was included in the Sixth International Workshop and Conference on Human Leucocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with CD57 (Funaro, 1997).

Specificity

Antibody recognizes Human CD57 antigen.

Immunogen

A pool of neuroblastoma cell lines

Application

Milli-Mark Anti-CD57-FITC Antibody, clone TB01 is an antibody against CD57 for use in FC.

Quality

Evaluated by flow cytometry using human PBMCs

Target description

110 kDa Observed

Physical form

Purified Mouse monoclonal IgM conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service