Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB170F

Sigma-Aldrich

Milli-Mark Anti-CD4 -FITC Antibody, clone MT310

clone MT310, Milli-Mark®, from mouse

Synonym(s):

T-cell surface antigen T4/Leu-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€336.00

€336.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 TESTS
€336.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€336.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

MT310, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD4(920)

General description

CD4 is a 55 kDa glycoprotein with four immunoglobulin-like external domains, a transmembrane domain and a highly conserved intracellular domain (Leong, 1999). CD4 acts as a coreceptor in MHC class II-restricted antigen-induced T-cell activation. Further, CD4 has a role in the regulation of T-B lymphocyte adhesion in the absence of antigen recognition, and in thymic differentiation (Piatier-Tonneau, 2002). CD4 is expressed on thymocyte subsets, it marks helper/inducer T cells, and it is expressed in 55-65% of mature peripheral T cells. Subpopulations of suppressor or cytotoxic T cells can be identified among CD4-positive T cells. CD4 is also expressed on monocytes/macrophages, Langerhans cells and other dendritic cells. CD4 is not expressed on B cells. Anti-CD4, MT310, was included in the Second International Workshop and Conference on Human Leucocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with the CD4 antigen (Miedema, 1986).

Specificity

Antibody recognizes Human CD4 antigen.

Application

Milli-Mark Anti-CD4 -FITC Antibody, clone MT310 is an antibody against CD4 for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human lymphocytes.

Target description

55 kDa (Leong, 1999)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 kappa conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service