Skip to Content
Merck
All Photos(1)

Key Documents

FCABS384F

Sigma-Aldrich

Milli-Mark® Anti-monomethyl-Histone H3 (Lys9)-FITC Antibody

Milli-Mark®, from rabbit

Synonym(s):

H3K9me1, Histone H3 (monomethyl K9), H3 histone family, member T, Histone 3, H3, Histone cluster 3, H3

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€286.00

€286.00


Estimated to ship on04 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 TESTS
€286.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€286.00


Estimated to ship on04 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Specificity

Broad species cross-reactivity is expected based on sequence homology.

Immunogen

Epitope: methylated lysine 9
KLH-conjugated, synthetic 2X-branched peptide corresponding to histone H3 monomethylated at lysine 9.

Application

Detect monomethyl-Histone H3 (Lys9) using this Milli-Mark Anti-monomethyl-Histone H3 (Lys9)-FITC Antibody validated for use in FC.
Research Category
Epigenetics & Nuclear Function

Epigenetics & Nuclear Function
Research Sub Category
Histone Modifying Proteins

Histones

Quality

Evaluated by flow cytometry using HeLa cells.

Target description

16 kDa Calculated

Physical form

Protein A purified
Purified rabbit polyclonal IgG conjugated to FITC in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
HeLa cells

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service