Skip to Content
Merck
All Photos(1)

Key Documents

EZIL6

Sigma-Aldrich

Human IL-6 ELISA Kit

detect IL-6 in 50 μL serum, plasma or cell culture supernatent

Synonym(s):

B-cell stimulatory factor 2, CTL differentiation factor, Hybridoma growth factor, IL6, Interferon beta-2, Interleukin-6

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€576.00
5 X 1 PKG
€2,730.00
10 X 1 PKG
€4,970.00

€576.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 PKG
€576.00
5 X 1 PKG
€2,730.00
10 X 1 PKG
€4,970.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.77

€576.00


Estimated to ship on07 April 2025Details


Request a Bulk Order

Product Name

Human IL-6 Conferma® ELISA, quantification of human IL-6 in serum, plasma, or culture media samples

Quality Level

species reactivity

human

parameter

50 μL sample volume (4hr assay)

assay range

linearity: 105-116%
(serum)

linearity: 88-105%
(plasma)

inter-assay cv: <15%
intra-assay cv: <5%
standard curve range: 1.17-300 pg/mL
accuracy: 91-109%

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma (K2 EDTA)
sample type cell culture supernatant

UniProt accession no.

application(s)

research use

detection method

colorimetric (450nm/590nm)

Gene Information

human ... IL6(3569)

General description

This Human Interleukin-6 (IL-6) Conferma® ELISA kit is used for the quantification of Human IL-6 in serum, plasma, or culture media samples. Human IL-6 is a soluble protein cytokine most commonly associated with the acute inflammatory response to infection or tissue damage.

This is a 96 well format sandwich ELISA which uses our Conferma® process to ensure a high degree of lot to lot consistency. The process involves the physiochemical characterization of the ELISAs critical reagents lot to lot, with finished kits being evaluated using a sample library to bridge between lots. For detailed information on the process and a Verification report for this assay please click hereConferma™ Sandwich ELISAs

Entrez Gene Summary:
This gene encodes a cytokine that functions in inflammation and the maturation of B cells. In addition, the encoded protein has been shown to be an endogenous pyrogen capable of inducing fever in people with autoimmune diseases or infections.

UniProt Summary:
  • Function: Cytokine with a wide variety of biological functions in immunity, tissue regeneration, and metabolism.
  • Size: 212 amino acids; 23,718 Da
  • Subunit: Component of a hexamer of two molecules each of IL6, IL6R and IL6ST.
  • Subcellular location: Extracellular region or secreted
  • Disease: Rheumatoid arthritis systemic juvenile (RASJ). A IL6 promoter polymorphism is associated with a lifetime risk of development of Kaposi sarcoma in HIV-infected men.
  • Similarity: Belongs to the IL-6 superfamily.

Application

One kit is sufficient to measure 37 samples in duplicate

Research Category:
  • Cytokines
  • Inflammation & Immunology
  • Immune Response
  • Metabolic Disease

Analysis Note

3 hour 45 minute assay, room temperature, 50 μL sample size (serum, plasma, or tissue culture medium)

Legal Information

CONFERMA is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For research use only. Not for use in diagnostic procedures.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the Conferma® ELISA development and manufacturing methods that provide strong sample detection and long-term assay and lot consistency, giving you confidence in your research.

Protocols

This article offers 4 popular ELISA protocols: Sandwich ELISA protocol, Phosphorylation Assay Procedure, EIA Assay Procedure, & Cell-based Assay Procedure.

Related Content

Learn how immunoassays are used in cosmetics and personal care research to discover potential harmful effects of cosmetics products, and assess markers of inflammation, sensitization, aging, and tissue regeneration among others.

Find immunoassays to explore neuroimmunology and neuroinflammation cytokines and neurodegenerative disease biomarkers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service