Skip to Content
Merck
All Photos(1)

Documents

EZFREAD01

Millipore

EZ-Fluo Reader

Synonym(s):

Detection instrument

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NA.74

AC/DC input

100 - 240 V, 50 - 60 Hz

description

For rapid detection of microbial contamination

manufacturer/tradename

EZ-Fluo

parameter

at 15 - 40 °C
4.4 kg

H × W × L

12.5 cm × 14.2 cm × 24.9 cm

application(s)

environmental
food and beverages

compatibility

EZ-Fluo Rapid Microbial Detection System

General description

The EZ-Fluo reader is part of the EZ-Fluo Rapid detection system. It is a fluorescent-based technology designed for fast and quantitative detection of microorganisms over a broad range of filterable samples. Fluorescent micro-colonies can be counted directly through the viewing window of the reader.

Application

The EZ-Fluo reader is a Rapid & non-destructive system for microbial detection in filterable samples.

Legal Information

EZ-FLUO is a trademark of Merck KGaA, Darmstadt, Germany

related product


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service