Skip to Content
Merck
All Photos(1)

Key Documents

EX0510

Supelco

Ethylenediamine

Synonym(s):

1,2-Ethanediamine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12191502
vapor pressure:
12 hPa ( 20 °C)

vapor pressure

12 hPa ( 20 °C)

Quality Level

Assay

≥98%

form

liquid

pH

12.2 (20 °C, 100 g/L in H2O)

mp

11 °C

transition temp

flash point 38 °C

storage temp.

room temp

Application

  • Novel Water-Soluble Ruthenium-Arene Ethylenediamine Schiff Base (RAES) Organometallic: This study explores the dual antimicrobial-anticancer potentials, hydrolysis, and DNA/BSA binding affinity of RAES, which is insightful for both chemical analysis and potential therapeutic applications (Zahirovic et al., 2024).
  • Enhanced adsorption and removal of Cd(II) from aqueous solution by amino-functionalized ZIF-8: This research focuses on the utilization of ethylenediamine-modified frameworks for environmental cleanup, specifically for the adsorption of cadmium from water, highlighting the adaptability of ethylenediamine in environmental applications (Khosravi et al., 2024).

Analysis Note

Assay:98% min
Form:Clear liquid, may form carbonates

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

100.4 °F - closed cup

Flash Point(C)

38 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service