EQ70XXPKT1
Milli-Q® EQ 7008/16 Purification Cartridge Kit
This product is replaced with EQ70XXPKIT. Search to Explore!
Select a Size
0800-14747.
This item is not able to be purchased as a guest user. For further information, please contact us atSelect a Size
About This Item
0800-14747.
This item is not able to be purchased as a guest user. For further information, please contact us atRecommended Products
material
carbon block
carbon filter (activated)
filter (Pleated)
filter (soda lime)
media (IQnano™ ion exchange)
media (Jetpore® mixed-bed ion exchange)
Quality Level
feature
Kit contains IPAK Gard® Pretreatment Pack, IPAK Meta®, Polishing CartridgeIPAK Quanta® Polishing Cartridge and a Tank Vent Filter.
manufacturer/tradename
Milli-Q®
compatibility
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
shipped in
ambient
General description
- IPAK Gard® pretreatment pack efficiently removes colloids, particles and free chlorine from tap water to protect the reverse osmosis (RO) membrane and improve system performance.
- IPAK Meta® and IPAK Quanta® polishing cartridges contain innovative Jetpore® mixed-bed ion-exchange resin and IQnano™ ion-exchange media, which achieve ion removal down to trace levels. In addition, high-grade synthetic activated carbon targets trace organic contaminants.
- The Tank Vent Filter, redesigned for seamless integration, provides improved protection against airborne contaminants.
Application
Features and Benefits
- Activated carbon to protect against free chlorine.
- Depth filter to protect against particulates and colloids.
Tank Vent Filter contains:
- Activated carbon to remove volatile organics.
- Soda lime to remove CO2.
- Membrane filter to prevent particulates and bacteria from entering the tank.
IPAK Meta® cartridge contains:
- Jetpore® mixed-bed ion-exchange resin and IQnano™ ion-exchange media to remove ions down to trace levels.
IPAK Quanta® cartridge contains:
- Innovative IQnano™ ion-exchange media to remove ions down to trace levels.
- High-grade synthetic activated carbon targets trace organic contaminants.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service