Skip to Content
Merck
All Photos(1)

Key Documents

ECM670-1

Sigma-Aldrich

QCM Gelatin Invadopodia Assay (Green)

Sign Into View Organizational & Contract Pricing

Select a Size

ECM670-1
€622.00

€622.00


Estimated to ship on10 April 2025Details



Select a Size

Change View
ECM670-1
€622.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

€622.00


Estimated to ship on10 April 2025Details


Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service