Skip to Content
Merck
All Photos(1)

Key Documents

ECM504

Sigma-Aldrich

QCM Chemotaxis Cell Migration Assay, 24-well (3µm), colorimetric

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€596.00

€596.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
€596.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.76

€596.00


Please contact Customer Service for Availability

species reactivity

all

Quality Level

technique(s)

activity assay: suitable

Application

The Millipore 3 µm QCM Chemotaxis Assay 24-well-colorimetric is performed in a Migration Chamber, based on the Boyden chamber principle. The 3 µm pore size of this assay′s Boyden chambers is appropriate for studying leukocyte migration. The quantitative nature of this assay is useful for screening of pharmacological agents. Each kit provides sufficient materials for the evaluation of 24 samples.

The Millipore 3 µm QCM Chemotaxis Assay 24-well-colorimetric is intended for research use only; not for diagnostic applications.

Packaging

24 wells

Components

Sterile 3 µm 24-well Cell Migration Plate Assembly: (Part No. 2005709) Two 24-well plates with 12 inserts per plate (24 inserts total/kit). WST-1 Reagent: (Part No. CS201370) One vial. Electro Coupling Solution: (Part No. CS201382) 5 ml. Forceps: (Part No. 10203) One each.

Storage and Stability

The two Sterile 3 µm 24-well Cell migration plate assembly and forceps are stored at 2°-8°C. WST-1 reagent and Electro Coupling Solution are stored at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service