ECM220
QCM Laminin Migration Assay (24-well, colorimetric)
Select a Size
€681.00
Select a Size
About This Item
€681.00
Recommended Products
manufacturer/tradename
Chemicon®
QCM
Quality Level
technique(s)
cell based assay: suitable
detection method
colorimetric
fluorometric
General description
Cell migration may be evaluated using several different methods; the most widely accepted being the Boyden Chamber assay. The Boyden Chamber system uses a two-chamber plate model in which a porous membrane provides an interface between two chambers. Cells are seeded in the upper chamber and chemoattractants placed in the lower chamber. Cells in the upper chamber migrate toward the chemoattractants by passing through the porous membrane to the lower chamber. Migratory cells are stained and quantified. Most cells are sized from 30 to 50 μm can migrate through 3 to 10 μm pore.
Application
Cell Structure
Laminin protein is coated on the bottom of each chamber′s porous membrane as the driver for cell migration. A similarly BSA-coated control chamber(s) provides an appropriate migration control. Cells that migrate toward laminin are stained using crystal violet, a nucleic dye, extracted from the membrane surface, and quantified on a microplate reader using spectrophotometry.
Each kit provides sufficient materials for the evaluation of 24 samples.
Packaging
Components
1. 24-well Cell Migration Plate Assembly - 5 μm: (Part No. 2005708) Two 24-well plates, each containing 12- 5 μm pore inserts per plate.
2. BSA 30% solution: (Part No. CS203352) One vial containing 0.5 mL of a 30% solution.
3. Cell Stain Solution*: (Part No. 90144) One bottle.
4. Extraction Buffer: (Part No. 90145) One bottle.
5. 24-well Stain Receiver Plate: (Part No. PIMWS2450) One each.
6. 96-Well Stain Quantitation Plate: (Part No. 2005870) One each.
7. Swabs: 50 ea
8. Forceps: 1 pair
*Caution: Cell Stain Solution contains a small amount of crystal violet, which is toxic if swallowed or inhaled, and may cause irritation to the eyes, respiratory system, and skin. Handle with caution.
ECM220-2
1. Laminin: (Part No. CS202685) Two vials each containing 60 μg in solution.
Storage and Stability
- ECM220-1 kit components should be stored at 2° to 8°C. DO NOT FREEZE.
- ECM220-2 component must be stored at -20°C until needed.
Legal Information
Disclaimer
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service