Skip to Content
Merck
All Photos(1)

Key Documents

DX0838

Supelco

Dichloromethane

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Synonym(s):

Methylene Chloride, FM approval, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-838-9
grade:
reagent
technique(s):
HPLC: suitable
bp:
39.8-40 °C (lit.)
vapor pressure:
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
Pricing and availability is not currently available.

grade

reagent

Quality Level

Agency

meets USP/NF

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

1223 °F

expl. lim.

22 %

technique(s)

HPLC: suitable

dilution

(HPLC: suitable)

impurities

≤0.0003 meq/g Titrable acid
≤0.02% Water (H2O)

evapn. residue

≤5 ppm

color

APHA: ≤10

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

solubility

20 g/L

density

1.325 g/mL at 25 °C (lit.)

UV cutoff

≤235 nm

UV absorption

λ: 235 nm Amax: ≤1.00
λ: 240 nm Amax: ≤0.35
λ: 250 nm Amax: ≤0.10
λ: 260 nm Amax: ≤0.04
λ: 270 nm Amax: ≤0.01
λ: 340-400 nm Amax: ≤0.01

shipped in

ambient

storage temp.

room temp

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DX0838P-1 is packaged in poly coated glass bottles
Stabilized with an alkene.
HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

AppearanceClear liquid, free from particulatesAssay (GC)99.8% minColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFree halogensTo pass testInfrared SpectrumConforms to standardTitrable acid0.0003meq/g maxUV Abs. at 235 nm1.00AU maxUV Abs. at 240 nm0.35AU maxUV Abs. at 250 nm0.10AU maxUV Abs. at 260 nm0.04AU maxUV Abs. at 270 nm0.01AU maxUV Abs. at 340 - 4000.01AU maxUV Cut-off235nm maxWater (H2O)0.02% max
Evaporation Number: 1.9

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service