Skip to Content
Merck
All Photos(1)

Key Documents

D01003S01

Millipore

Clarigard® Cartridge Filter

nominal pore size 1.0 μm, cartridge nominal length × diam. 30 in. (757.0-765.0 mm) × 2.75 in. (70 mm)

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€281.00

€281.00


Estimated to ship on17 September 2025Details



Select a Size

Change View
1 PKG
€281.00

About This Item

UNSPSC Code:
23151806

€281.00


Estimated to ship on17 September 2025Details


material

polypropylene filter

Quality Level

feature

hydrophilic

manufacturer/tradename

Clarigard®

parameter

4.8 bar max. differential pressure (70 psid) at 20 °C

cartridge nominal length × diam.

30 in. (757.0-765.0 mm) × 2.75 in. (70 mm)

impurities

≤0.25 EU/mL bacterial endotoxins (LAL test, aqueous extraction)

gravimetric extractables

≤10 mg

pore size

1.0 μm nominal pore size
1.0 μm pore size

input

sample type liquid

cartridge code

Code 0 (2-222; O-Ring)

Looking for similar products? Visit Product Comparison Guide

General description

Clarigard® filters 99.99% retention characteristic makes them ideal for the protection of critical downstream process steps such as membrane filters or chromatography columns. The graded-density depth structure of Clarigard® filter media provides maximum filtration capacity, and the polypropylene construction offers low extractable levels and broad chemical compatibility.

Preparation Note

Sterilization: May be autoclaved for 10 cycles of 30 minutes at 126 °C or steam sterilized for 10 cycles of 30 minutes at 126 °C or hot water sanitized at 80 °C maximum for 30 minutes.

Analysis Note

Gravimetric Extractables: After autoclaving and a 24-hour soak in ASTM® Type 1 reagent grade water at controlled room temperature, meets the USP Oxidizable Substances Test requirements of <10 mg.

Other Notes

Directions for Use:
  • Organism Retention: 99.99% retention
  • Mode of Action: Filtration (nominal)
  • Application: BioProcessing
  • Intended Use: Bioborden reduction
  • Instructions for Use: Please refer to the Polygard® and Clarigard® Cartidges user guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
CLARIGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
POLYGARD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service