max. pressure at 3.15 bar and 25 °C 45 °C max. temp.
L
850 mm
pore size
0.45 μm (Canister membrane)
input
liquid
application(s)
pharmaceutical sterility testing
compatibility
for use with Steritest® systems
General description
The SteriSecure™ Device for ampoules and bags with 3 canisters is designed for sterility testing of large volume parenterals and synthetic drugs. The SteriSecure™ Device for liquids in ampoules or collapsible bags includes a single needle for easy access to ampoules and a separate vent needle. The closed system minimizes false positives and offers the highest level of quality and reliability. This device ensures that pharmaceutical products are never exposed to the environment during the testing process. This membrane provides an optimal filtration flow rate for standard products. This test system offers an optimized and fully regulatory compliant testing process when used with the Steritest® Symbio pump, specific accessories, high-quality culture media and rinsing fluids.
Application
The SteriSecure™ Device for ampoules and bags with 3 canisters is used during the testing of pharmaceutical products in ampoules or collapsible bags for streamlining the workflow and increasing safety during sterility testing.
Legal Information
STERISECURE is a trademark of Merck KGaA, Darmstadt, Germany
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.