Skip to Content
Merck
All Photos(1)

Documents

CHR104

Sigma-Aldrich

DNA extraction kit for ChIC/CUT&RUN

12 RXNs

Synonym(s):

DNA extraction kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.52

General description

The DNA extraction by using a spin column is recommended for recovery of large protein–DNA complexes such as nucleosomes in ChIC/CUT&RUN experiment. If you are processing recovery of small protein-DNA complexes such as transcription factors, the alternative DNA extraction (phenol chloroform extraction, section VII) is recommended.For details, please refer to the manual of ChIC (Chromatin Immuno-Cleavage) Kits (Part #s: CHR100-102).

Application

The DNA extraction kit utilizes column purification and is recommended for large protein-DNA complexes such as nucleosomes. For quantitative recovery of < 80bp fragments, the alternative extraction method is recommended in the ChIC kit user guide.

Other Notes

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service