CDRM6HN05
Lynx® CDR
Lynx® CDR Male 3/4 Inch Hose Barb Reconnectable Connector - 5pack
Synonym(s):
Multiple aseptic connections, disconnections and reconnections (up to 6 times)
Select a Size
€1,936.00
Estimated to ship on09 May 2025Details
Select a Size
About This Item
€1,936.00
Estimated to ship on09 May 2025Details
Recommended Products
material
316 stainless steel spring(s) (electropolished)
polyethersulfone membrane (for vent)
polysulfone flow path (glass filled polysulfone USP Class VI, ADCF)
polysulfone housing (outer, sterility chamber)
polysulfone valve body (glass filled polysulfone USP Class VI, ADCF)
silicone seal (USP Class VI, ADCF or EMA/410/01 compliant)
Quality Level
sterility
non-sterile
product line
Lynx®
parameter
0.7 bar max. pressure (10 psi) (connect/disconnect pressure)
2-40 °C operating temperature (36-104 °F)
4.1 bar max. pressure (60 psi) (operating pressure)
H
13.7 cm (5.4 in.)
L
25.7 cm (10.1 in.)
W
8.4 cm (3.3 in.)
fitting
3/4 in. inlet hose barb
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
What is the Emprove® Program?
The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:
Free of charge - Sign in to download
Free of charge - Sign in to download
Available by subscription or for a fee
Available by subscription or for a fee
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service