Skip to Content
Merck
All Photos(1)

Key Documents

CC1059

Sigma-Aldrich

MMP-7, human, recombinant active form

Synonym(s):

Matrilysin

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€526.00

€526.00


Estimated to ship on11 February 2025Details



Select a Size

Change View
10 μG
€526.00

About This Item

UNSPSC Code:
12352204
eCl@ss:
32160405
NACRES:
NA.41

€526.00


Estimated to ship on11 February 2025Details


biological source

human

Quality Level

form

liquid

specific activity

5,000 units/mg protein

manufacturer/tradename

Chemicon®

concentration

0.1 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

MOLECULAR WT:

19,130 calculated from amino acid sequence and 20,000 from SDS-PAGE, followed by staining with CBB R-250.
Product Source: Recombinant E. Coli; prepared from recombinant human promatrilysin

Unit Definition

Specific Activity: One unit of enzyme activity is defined as the quantity required to digest 1 μg of Azocoll/min.at pH 7.5 and 37°C in the presence of 0.5 mM p-aminophenylmercuric acetate.

Physical form

Solution in 10 mM HEPES (pH 7.4)-5 mM CaCl2-0.15 M NaCl.

Storage and Stability

Maintain at -80ºC for up to one year.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tristen V Tellman et al.
International journal of molecular sciences, 22(6) (2021-04-04)
The Perlecan-Semaphorin 3A-Plexin A1-Neuropilin-1 (PSPN) Complex at the cell surface of prostate cancer (PCa) cells influences cell-cell cohesion and dyscohesion. We investigated matrix metalloproteinase-7/matrilysin (MMP-7)'s ability to digest components of the PSPN Complex in bone metastatic PCa cells using in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service