Skip to Content
Merck
All Photos(1)

Key Documents

CBL266

Sigma-Aldrich

Anti-Cytokeratin 1 Antibody, 10, clone LH1

clone LH1, Chemicon®, from mouse

Synonym(s):

Anti-CK1, Anti-EHK, Anti-EHK1, Anti-EPPK, Anti-K1, Anti-KRT1A, Anti-NEPPK

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€343.00

€343.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€343.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€343.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

LH1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KRT1(3848)

Specificity

This antibody reacts with the suprabasal cells of stratified squamous epithelia in human, murine and porcine species.

Immunogen

Keratin 1 and 10.

Application

Detect Cytokeratin 1 using this Anti-Cytokeratin 1 Antibody, 10, clone LH1 validated for use in WB, IH.
Research Category
Cell Structure
Research Sub Category
Cytokeratins
Western blotting.

IHC formalin fixed sections, HIER (Microwave citrate buffer pH6.0) produces some staining but pretreatment with type XXIV protease (0.5mg/ml) produces best results.

Optimal working dilutions must be determined by the end user.

Physical form

Format: Purified
The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular application.

Storage and Stability

For use within 1 month of purchase store at +4°C, for long term storage aliquot antibody into small volumes and store at -20°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service