Skip to Content
Merck
All Photos(1)

Key Documents

CBL1512F

Sigma-Aldrich

Anti-Integrin αM [CD11b] Antibody, clone OX-42, FITC conjugated

clone OX-42, Chemicon®, from mouse

Synonym(s):

CD11b

Sign Into View Organizational & Contract Pricing

Select a Size

100 ASSAYS
€408.00

€408.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

Select a Size

Change View
100 ASSAYS
€408.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€408.00


Estimated to ship on05 May 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody product type

primary antibodies

clone

OX-42, monoclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

Specificity

This antibody recognizes most macrophages (including resident peritoneal and activated macrophages), kupffer cells, but only about 35% of alveolar macrophages. The antibody also labels dendritic cells extensively, granulocytes and cells with the morphology of microglia in brain. The antibody precipitates three polypeptides of 150, 103 and 95 KDa. The antibody inhibits complement mediated rosettes and is probably the rat equivalent of the human receptor for IC3b called CR3.
FUSION PARTNER: NSO myeloma cell line.

Application

Flow cytometry

Immunohistochemical studies of rat fresh frozen tissue sections following either periodate-lysine-paraformaldehyde (PLP) fixation, or acetone. Works on very lightly PFA fixed, frozen tissues. (perfusion only 4% PFA 10-15′ no post-fix).

Optimal working dilutions must be determined by the end user.
This Anti-Integrin αM CD11b Antibody, clone OX-42, FITC conjugated is validated for use in FC, IH for the detection of Integrin αM [CD11b].

Linkage

Replaces: MAB1404F

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaesuk Lee et al.
International journal of stem cells, 12(1), 114-124 (2019-03-07)
Parkinson’s disease (PD) is a fatal and progressive degenerative disease of the nervous system. Until recently, its promising treatment and underlying mechanisms for neuronal death are poorly understood. This study was investigated to identify the molecular mechanism of neuronal death
Kyunghee Byun et al.
PloS one, 9(8), e104699-e104699 (2014-08-21)
Alcohol is a neurotoxic agent, since long-term heavy ingestion of alcohol can cause various neural diseases including fetal alcohol syndrome, cerebellar degeneracy and alcoholic dementia. However, the molecular mechanisms of alcohol-induced neurotoxicity are still poorly understood despite numerous studies. Thus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service