Skip to Content
Merck
All Photos(1)

Documents

BX1673

Supelco

2-Butanone

OmniSolv®

Synonym(s):

2-Butanone, Methyl Ethyl Ketone, Butanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-159-0

vapor pressure

105 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥99.5% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.05% Water (H2O)

evapn. residue

≤5 ppm

color

APHA: ≤10

pH

7 (20 °C)

bp

79.6 °C/1013 hPa

mp

-86 °C

transition temp

flash point -4 °C

solubility

292 g/L

density

0.805 g/cm3 at 20 °C

UV cutoff

≤329 nm

UV absorption

λ: 329 nm Amax: ≤1.00
λ: 335 nm Amax: ≤0.30
λ: 344 nm Amax: ≤0.05
λ: 350 nm Amax: ≤0.01
λ: 400 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

InChI

1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3

InChI key

ZWEHNKRNPOVVGH-UHFFFAOYSA-N

Related Categories

Application

BX1673P-1 is packaged in poly coated glass bottles
OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.5% minColor (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 329 nm1.00AU maxUV Abs. at 335 nm0.30AU maxUV Abs. at 344 nm0.05AU maxUV Abs. at 350 nm0.01AU maxUV Abs. at 400 nm0.005AU maxUV Cut-off329nm maxWater (H2O)0.05% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

30.2 °F - closed cup

Flash Point(C)

-1 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service