Skip to Content
Merck
All Photos(1)

Key Documents

BM-1E

Millipore

Auto2D® Electrode Chip

Auto2D® Electrophoresis Device

Synonym(s):

2D Electrophoresis, 2DE, Electrode, Electrode Chip, IEF, SDS-PAGE

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€1,520.00

€1,520.00


Estimated to ship on22 April 2025Details



Select a Size

Change View
1 PKG
€1,520.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

€1,520.00


Estimated to ship on22 April 2025Details


product line

Auto2D®

compatibility

Auto2D® Electrophoresis Device

storage temp.

room temp

General description

Electrode Chip is the essential part of the Auto2D® 2-D Electrophoresis Device when running the system with "Auto2D" mode.

Application

  • Differential proteomic analysis
  • Post-translational modification
  • Protein Isoforms
  • Western Blotting

Storage and Stability

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Legal Information

Auto2D is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Auto2D® automates difficult 2D electrophoresis methods for protein localization with high reproducibility in under two hours.

Protocols

The Auto2D® Automated 2-D Gel Electrophoresis Device video and detailed protocol provide reliable methods to prepare your samples and operate the Auto2D® Device.

Related Content

Auto2D® 2-D Gel Electrophoresis Device frequently asked questions for 2-D PAGE and additional 2DE protein separation workflow needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service