Skip to Content
Merck
All Photos(1)

Key Documents

BCYT1-33K-PXBK15

Millipore

MILLIPLEX® Bovine Cytokine/Chemokine Magnetic Bead Premixed 15-plex Panel 1 Space Saver (Bulk) Packaging - Immunology Multiplex Assay

Synonym(s):

Magnetic bead assay

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€3,470.00

€3,470.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

Select a Size

Change View
1 EACH
€3,470.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.47

€3,470.00


Product Availability: You will receive an order acknowledgement once the order has been entered in our system.

Request more information

species reactivity

bovine

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

Inflammation and immune cell response are key to maintaining homeostasis. Inflammation plays a central role in the etiology of many immunologically related diseases, such as infectious disease, osteoarthritis and respiratory disease, as well as neurologic, metabolic and cardiovascular disease and even cancer. However, until now the ability to measure levels of cytokines and chemokines in bovine samples has been limited primarily to ELISAs. Researchers can now measure multiple inflammatory biomarkers simultaneously using our MILLIPLEX® Bovine Cytokine/Chemokine Panel 1, based on the Luminex® xMAP technology.The MILLIPLEX® Bovine Cytokine/Chemokine Magnetic Bead Premixed 15-plex Panel 1 is a 15-plex kit to be used for the simultaneous quantification of all of the following analytes in serum, plasma, and tissue culture supernatant samples: IFNγ, IL-1α, IL-1β, IL-4, IL-6, IL-8 (CXCL8), IL-10, IL-17A, IL-36RA (IL-1F5), IP-10 (CXCL10), MCP-1 (CCL2), MIP-1α (CCL3), MIP-1β (CCL4), TNFα, and VEGF-A.MILLIPLEX® offers the broadest selection of analytes across a wide range of disease states and species. Once the analytes of interest have been identified, you can rely on the quality that we build into each kit to produce results you can trust. In addition to the assay characteristics listed in the protocol, other performance criteria evaluated during the validation process include: cross-reactivity, dilution linearity, kit stability, and sample behavior (e.g. detectability and stability). Each panel and kit meets stringent manufacturing criteria to ensure batch-to-batch reproducibility.Each MILLIPLEX® panel and kit includes:Quality controls (QCs) provided to qualify assay performanceComparison of standard (calibrator) and QC lots to a reference lot to ensure lot-to-lot consistencyOptimized serum matrix to mimic native analyte environmentDetection antibody cocktails designed to yield consistent analyte profiles within panelThe MILLIPLEX® Bovine Cytokine/Chemokine Magnetic Bead Panel 1 is part of the most versatile system available for cytokine and chemokine research. From our single to multiplex biomarker solutions, we partner with you to design, develop, analytically validate and build the most comprehensive library available for protein detection and quantitation.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

The Bovine Cytokine/Chemokine bead-based multiplex cytokine panel, using the Luminex® xMAP technology, enables the simultaneous analysis of 15 cytokine and chemokine biomarkers in bovine serum, plasma and cell culture samples.This is an overnight incubation assay (or you may incubate at room temperature for 2 hours with shaking).A maximum of 25 μL per well 1:2 diluted serum or plasma can be used. Tissue culture or other media may be run neat or appropriately diluted in control medium.This kit may be used for the analysis of all the Bovine Cytokine/Chemokine Panel 1 analytes in serum, plasma or tissue/cell/tumor culture supernatant.Analytes available:IFNγ;IL-1α;IL-1β;IL-4;IL-6;IL-8;IL-10;IL-17A;MIP-1α;IL-36RA;IP-10;MCP-1;MIP-1β;TNFα;VEGF-A

Packaging

96-well plate

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2

Target Organs

Respiratory Tract

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service