Skip to Content
Merck
All Photos(1)

Key Documents

AX1697C

Supelco

Coulomat C

AQUASTAR®

Synonym(s):

Coulometric Cathode Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115601

Quality Level

form

liquid

transition temp

flash point 16 °C

shipped in

ambient

storage temp.

room temp

Application

AQUASTAR
AX1697C-1 is packaged in 20/400 glass bottle
AX1697C-30 is packaged in 38/430 poly-coated glass bottle

Analysis Note

Color: Colorless to light yellow
Form: Liquid
Performance: To pass test
Water (H2O): 1-50ppm

Legal Information

AQUASTAR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Ozone 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT RE 2 Oral - STOT SE 1

Target Organs

Eyes,Central nervous system, Kidney,Liver,Blood, Liver,Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service