Skip to Content
Merck
All Photos(1)

Key Documents

AX0745

Supelco

Aluminum Sulfate, Octadecahydrate

Synonym(s):

Aluminum Sulfate, Octadecahydrate, Cake alum,Patent alum, Alum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2(SO4)3 · 14-18H2O
CAS Number:
UNSPSC Code:
12352302
EC Index Number:
233-135-0
form:
solid
solubility:
364 g/L
Pricing and availability is not currently available.

form

solid

Quality Level

concentration

54.0-59.0% Al2(SO4)3

pH

2.5-4.0 (20 °C, 20 g/L in H2O)

mp

90 °C (decomposition)

solubility

364 g/L

cation traces

As: ≤0.001%
Fe: ≤0.005%
heavy metals (as Pb): ≤0.005%

shipped in

ambient

storage temp.

room temp

InChI

1S/2Al.3H2O4S.9H2O/c;;3*1-5(2,3)4;;;;;;;;;/h;;3*(H2,1,2,3,4);9*1H2/q2*+3;;;;;;;;;;;;/p-6

InChI key

KFCWRPFNNIEZRY-UHFFFAOYSA-H

Analysis Note

Arsenic (As): 0.001% max
Assay (as Al2(SO4)3): 54.0-59.0%
Heavy metals (as Pb): 0.005% max
Iron (Fe): 0.005% max

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service