Skip to Content
Merck
All Photos(2)

Key Documents

APT225

Sigma-Aldrich

ssDNA Apoptosis ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000

species reactivity

pig, human, rat, horse, hamster, mouse, guinea pig, rabbit, canine, nonhuman primates

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

General description

This procedure is based on the selective denaturation of DNA in apoptotic cells by formamide, and detection of denatured DNA with monoclonal antibody to single-stranded DNA (ssDNA). Formamide is a gentle agent that denatures DNA in apoptotic cells, but not in necrotic cells or in the cells with DNA breaks in the absence of apoptosis (Frankfurt & Krishan 2001). The sensitivity of DNA in apoptotic cells to formamide is not related to DNA breaks, but rather reflects changes in chromatin associated with apoptosis, such as chromatin condensation and digestion of proteins stabilizing DNA. The assay includes attachment of cells to 96-well plates, treatment of attached cells with formamide, and staining of ssDNA in apoptotic cells with a mixture of primary antibody and peroxidase-labeled secondary antibody. The protocol based on the one-step detection of ssDNA with antibody mixture has higher sensitivity and lower number of steps than standard two-step immunostaining (Frankfurt & Krishan 2001). This mixture is included in the kit in a ready to use form.

Application:

The CHEMICON ssDNA Apoptosis ELISA Kit is a convenient, sensitive method for early detection of apoptosis. The assay can be performed in two formats:

1. Cells are grown, treated with apoptosis inducing agents, stained and analyzed in the same microtiter plate. This assay is suitable for high throughput screening when induction of apoptosis is used as the endpoint of drug activity.

2. Cell suspensions obtained from cultures or tissues are transferred into a microtiter plate for staining and analysis. For this assay, cells could be fixed with methanol and stored before transfer into plates. Alternatively, fixation could be performed in microtiter plates after transfer of non-fixed cells.

Total assay time is 3-4 hours, which includes fixation and staining. Only one washing step is required.

The ssDNA Apoptosis ELISA is for research use only. Not for use in diagnostic or therapeutic procedures.

Application

Positive Control

To use the ssDNA positive control included in this kit:

1. Add 100 μL of ssDNA Positive Control solution per well and dry plate by floating in a 37ºC waterbath or in a 37ºC incubator or culture hood (fan on) with lid off, overnight or until very dry. WELLS MUST BE THROUGHLY DRY for ssDNA to adhere to the wells.

2. Rinse wells with PBS before use. Plate can be stored dry, covered until ready to use.

3. Proceed to step 8 of Assay Instructions below if positive control is done on a separate plate. If performing entire assay on one plate, proceed to sample preparation below, ignoring wells used for positive control UNTIL step 8.

Note: Absorbance reading between 1.5 and 2.8 indicates good assay sensitivity; lower values will still work, however it may indicate that some of the ssDNA has come off the plate. One can reapply ssDNA standard again, if necessary.
Research Category
Apoptosis & Cancer
Used to detect/quantify: ssDNA

Packaging

96 wells

Components

Antibody Mixture, Primary monoclonal to ssDNA and HRP-labeled goat anti-mouse IgM (Part No. 71278a): One 10 mL bottle, premixed (Ready to Use).

Formamide (Part No. 71278b): One 5 mL bottle (Ready to Use).

Single-stranded DNA Positive Control (Part No. 71278c): One 2 mL vial at 0.3 μg/mL.

Wash Buffer Concentrate (Part No. 90160): One 10 mL (10X) bottle of Concentrate.

ABTS Solution (Part No. 90161): One 12 mL bottle of a Ready to Use solution of 2,2′-AZINO-bis [3-ethylbenziazoline-6-sulfonic acid] in a proprietary buffer with enhancer.

Stop Solution (Part No. 90162): One 12 mL bottle of an HCl solution (Ready to Use).

Storage and Stability

Store kit materials at -20°C. Once the kit is opened, Antibody Mixture and ssDNA Positive Control should be thawed on ice, aliquoted and stored at -20ºC. Formamide, Wash Buffer, ABTS Solution and Stop Solution can be thawed and stored at 4°C.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birgit Jaschke et al.
Cardiovascular research, 68(3), 483-492 (2005-08-23)
Therapeutic strategies to provide local inhibition of mitogen mediated proliferation and migration of human coronary artery smooth muscle cells (CASMC) by means of drug-eluting stents have been shown to enable effective limitation of neointimal hyperplasia. However, currently available drug-eluting stents
Syh-Yuan Huang et al.
Journal of Asian natural products research, 11(5), 410-416 (2009-06-09)
A new dimeric phenylpropanoid namely podonaka A (1), along with the 13 known compounds including diterpenes (2 and 3), norditerpenes (4 and 5), benzenoids (6-10), steroids (11 and 12), chalcone (13), and megastigmane (14), was isolated from the EtOH extract
Cheng-Fang Yang et al.
International journal of molecular medicine, 41(4), 2306-2316 (2018-01-18)
The aim of the present study was to assess the molecular mechanism of ethanol‑induced oxidative stress‑mediated apoptosis in L‑02 liver cells in order to elucidate novel pathways associated with alcoholic liver disease. L‑02 cells were treated with 400 mM ethanol with
Jeffrey C Horowitz et al.
The Journal of biological chemistry, 279(2), 1359-1367 (2003-10-25)
Transforming growth factor-beta1 (TGF-beta1) is a multifunctional cytokine involved in differentiation, growth, and survival of mesenchymal cells while inhibiting growth/survival of most other cell types. The mechanism(s) of pro-survival signaling by TGF-beta1 in mesenchymal cells is unclear. In this report
Brian Bauereis et al.
Neuroscience letters, 488(1), 11-16 (2010-11-09)
Previous studies in Parkinson's disease (PD) models suggest that early events along the path to neurodegeneration involve activation of the ubiquitin-proteasome system (UPS), endoplasmic reticulum-associated degradation (ERAD), and the unfolded protein response (UPR) pathways, in both the sporadic and familial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service