Recommended Products
species reactivity
mammals
technique(s)
activity assay: suitable
General description
Internucleosomal DNA fragmentation is a hallmark of apoptosis in mammalian cells. Chemicon′s Quick Apoptotic DNA Ladder Detection Kit provides a fast and easy means for detection of DNA fragmentation in apoptotic cells. Unlike other similar kits which require 1-2 days to finish the procedure, the new detection method requires less than 90 minutes to prepare DNA in a single tube, without the need for extraction or the use of columns. DNA fragmentation can be easily visualized by agarose gel electrophoreses. The new procedure increases recovery of small fragmented DNA and therefore improves the sensitivity of the assay.
Components
TE Lysis Buffer - 2 mL. Enzyme A Solution - 0.25 mL. Enzyme B* - 1 vial Ammonium Acetate Solution - 0.25 mL. DNA Suspension Buffer - 2 mL. Note: Dissolve Enzyme B with 275 μL TE Lysis Buffer and mix well before use. The unused Enzyme B solution should be frozen at -70°C.
Storage and Stability
We recommend storing reagents at -20°C upon arrival. Frequent freeze-thawing is not recommended. The performance of this product is guaranteed for 6 months.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
10 - Combustible liquids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service