Skip to Content
Merck
All Photos(1)

Key Documents

AP512

Sigma-Aldrich

Sheep Anti-Human IgM, (Fc) Antibody

Chemicon®, from sheep

Synonym(s):

Ig mu chain C region

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€266.00

€266.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 μG
€266.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€266.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

biological source

sheep

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Immunoglobin M (IgM) antibody is prominent in early immune responses to most antigens and is the predominant response in certain antibody responses such as "natural" blood group antibodies. IgM is the major immunoglobin expressed on surface B lymphocytes. Most of the chronic lymphocytic leukemias, non Hodgkin lymphomas and Burkitt′s lymphoma bear surface IgM, whereas plasmocytes from Waldenström′s disease bear intracytoplasmic IgM.

Specificity

No reactivity with IgG or IgA. May react with IgM of other species with low specificity.
This antibody reacts with human IgM, (Fc) as demonstrated by ELISA.

Immunogen

Epitope: IgM, (Fc)
Human anti-Human IgM (myeloma) Fc5u fragment

Application

Research Category
Secondary & Control Antibodies
Research Sub Category
Isotype Control Antibodies
Sheep anti-Human IgM, (Fc) Antibody is an antibody against Human IgM, (Fc) for use in ELISA.

Quality

Evaluated by ELISA against Human IgG, Human IgG Fab Fragment, Human IgG Fc, Human IgM Fc, Human IgM (myeloma) Fab m fragment, Human IgA, Mouse IgM, Rat IgM whole molecule, Chicken IgM whole molecule, Goat IgM whole molecule, Guinea Pig IgM whole molecule, Hamster IgM whole molecule, and Rabbit IgM whole molecule.

ELISA Analysis: A representative lot detected Sheep Anti-Human IgM, (Fc) in ELISA.

Target description

50 kDa calculated

Physical form

Depletion
Format: Purified
Purified sheep polyclonal in PBS containing 0.05% sodium azide. Depleted against rabbit IgM and human IgG.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human IgG, Human IgG Fab Fragment, Human IgG Fc, Human IgM Fc, Human IgM (myeloma) Fab m fragment, Human IgA, Mouse IgM, Rat IgM whole molecule, Chicken IgM whole molecule, Goat IgM whole molecule, Guinea Pig IgM whole molecule, Hamster IgM whole molecule, and Rabbit IgM whole molecule

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service