Skip to Content
Merck
All Photos(1)

Key Documents

AP193B

Sigma-Aldrich

Donkey Anti-Guinea Pig IgG Antibody, biotin conjugate, Species Adsorbed

Chemicon®, from donkey

Sign Into View Organizational & Contract Pricing

Select a Size

500 μL
€347.00

€347.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
500 μL
€347.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€347.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

donkey

Quality Level

conjugate

biotin conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

guinea pig

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Donkey anti-Guinea Pig IgG Antibody, biotin conjugate, Species Adsorbed is an antibody against Donkey Guinea Pig IgG for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun-Bin Yin et al.
The Journal of clinical investigation, 130(12), 6555-6570 (2020-08-26)
The dorsal medial prefrontal cortex (dmPFC) has been recognized as a key cortical area for nociceptive modulation. However, the underlying neural pathway and the function of specific cell types remain largely unclear. Here, we show that lesions in the dmPFC
Brian H Kim et al.
Cells, 10(4) (2021-05-01)
Recent studies of cerebral hypoxia-ischemia (HI) have highlighted slowly progressive neurodegeneration whose mechanisms remain elusive, but if blocked, could considerably improve long-term neurological function. We previously established that the cytokine transforming growth factor (TGF)β1 is highly elevated following HI and
Kengo Nomura et al.
Neuron, 106(5), 816-829 (2020-04-02)
Sodium taste regulates salt intake. The amiloride-sensitive epithelial sodium channel (ENaC) is the Na+ sensor in taste cells mediating attraction to sodium salts. However, cells and intracellular signaling underlying sodium taste in taste buds remain long-standing enigmas. Here, we show
Xiao-Na Wu et al.
Neurochemistry international, 82, 1-9 (2015-02-11)
Endomorphin-2 (EM2) and Substance P (SP) exert suppressive and facilitative influences upon nociception, respectively. Although EM2 and SP were often co-expressed in single neurons in dorsal root ganglion (DRG), it is still unknown if and how the nociception-suppressive influences of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service