Skip to Content
Merck
All Photos(1)

Documents

AP127F

Sigma-Aldrich

Goat Anti-Mouse IgG Antibody, Fc, FITC conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Goat anti-Mouse IgG Antibody, Fc, FITC conjugate is an antibody against Mouse IgG for use in IF.
Research Category
Secondary & Control Antibodies
Research Sub Category
Fragment Specific Secondary Antibodies

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lian Zhou et al.
Molecular medicine reports, 25(1) (2021-11-19)
The cellular resistance of tumors is a major obstacle for successful tumor therapy. Cluster of differentiation (CD)133 plays an important role in the regulation of drug resistance in gastric and colon cancers. However, its effect on chemotherapeutic sensitivity in adenoid cystic
Noriaki Koyama et al.
Life sciences, 89(19-20), 741-747 (2011-10-01)
Mesenchymal stem cells are an interesting source of material for regenerative medicine. The present study aimed at characterizing the phenotype and differentiation potential of adherent synovial fluid-derived cells from temporomandibular joint (TMJ) disorder patients. Synovial fluid collection takes place during
Chenzhu Zhang et al.
Frontiers in pharmacology, 13, 866077-866077 (2022-05-17)
Background: A significant proportion of lupus nephritis patients develop chronic kidney disease (CKD) and progressive kidney fibrosis, for which there is no specific treatment. We previously reported that mycophenolate or rapamycin monotherapy showed comparable efficacy in suppressing kidney fibrosis in
Guang Song et al.
Genomics, proteomics & bioinformatics, 19(1), 108-122 (2021-02-22)
The Zika virus (ZIKV) and dengue virus (DENV) flaviviruses exhibit similar replicative processes but have distinct clinical outcomes. A systematic understanding of virus-host protein-protein interaction networks can reveal cellular pathways critical to viral replication and disease pathogenesis. Here we employed
Donghwa Kim et al.
The Journal of biological chemistry, 291(34), 17616-17628 (2016-06-28)
Bitter taste receptors (TAS2Rs) are G-protein-coupled receptors now recognized to be expressed on extraoral cells, including airway smooth muscle (ASM) where they evoke relaxation. TAS2Rs are difficult to express in heterologous systems, with most receptors being trapped intracellularly. We find

Related Content

Step-by-step protocol for generating apical-out human gut organoids for microbiome, ADME/Tox, viral and gastrointestinal related disease research. See the complete organoid culture protocol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service