Skip to Content
Merck
All Photos(1)

Key Documents

AP120R

Sigma-Aldrich

Goat Anti-Human IgA Antibody, IgG, and IgM, Rhodamine conjugate

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
€188.00

€188.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

Select a Size

Change View
2 MG
€188.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.47

€188.00


Estimated to ship on28 April 2025Details


Request a Bulk Order

biological source

goat

Quality Level

conjugate

TRITC conjugate
rhodamine conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunofluorescence: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Goat anti-Human IgA Antibody, IgG & IgM, Rhodamine conjugate is an antibody against Human IgA for use in IF.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junying Chen et al.
Bioengineered, 12(2), 10723-10733 (2021-11-10)
RAS p21 protein activator 4 (RASA4) has been recognized as a Ca2+-promoted Ras-MAPK pathway suppressor that inhibits tumor growth. However, the role of RASA4 in cervical squamous cell carcinoma (CESC) remains unclear. The mRNA levels of RASA4 were analyzed using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service