Skip to Content
Merck
All Photos(1)

Key Documents

AP106A

Sigma-Aldrich

Rabbit Anti-Goat IgG Antibody, Alkaline Phosphatase conjugate

Chemicon®, from rabbit

Synonym(s):

Rabbit anti-Goat antibody

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€380.00

€380.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
€380.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

€380.00


Estimated to ship on25 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

F(ab′)2 fragment of affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

goat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

Application

Rabbit anti-Goat IgG Antibody, Alkaline Phosphatase conjugate is an antibody against Goat IgG for use in ELISA & WB.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT RE 2 Oral

Target Organs

Brain

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Runzhao Guo et al.
Cytoskeleton (Hoboken, N.J.), 79(9-11), 94-104 (2022-09-03)
Microtubule stability and dynamics regulations are essential for vital cellular processes, such as microtubule-dependent axonal transport. Dynamin is involved in many membrane fission events, such as clathrin-mediated endocytosis. The ubiquitously expressed dynamin-2 has been reported to regulate microtubule stability. However
Guang-Jer Wu et al.
BMC cancer, 16, 136-136 (2016-02-26)
Increased expression of METCAM/MUC18, a trans-membrane cell adhesion molecule in the Ig-like gene superfamily, has been associated with the malignant progression of epithelial ovarian carcinomas. To investigate if this is a fortuitous correlation or if METCAM/MUC18 actually plays a role

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service