Skip to Content
Merck
All Photos(1)

Key Documents

AP1062

Sigma-Aldrich

PhosphoDetect Anti-PDH-E1α (pSer²⁹³) Rabbit pAb

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€596.00

€596.00


Estimated to ship on05 November 2025Details


Request a Bulk Order

Select a Size

Change View
50 μL
€596.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

€596.00


Estimated to ship on05 November 2025Details


Request a Bulk Order

Quality Level

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service