Skip to Content
Merck
All Photos(1)

Key Documents

AG383

Sigma-Aldrich

Capsaicin Receptor Control Peptide for AB5889

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€192.00

€192.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€192.00

About This Item

UNSPSC Code:
51111800
eCl@ss:
32160405
NACRES:
NA.41

€192.00


Estimated to ship on17 April 2025Details


Request a Bulk Order

concentration

1 mg/mL

Quality Level

technique(s)

inhibition assay: suitable

UniProt accession no.

Gene Information

General description

FUNCTION: SwissProt: Q8NER1 # Receptor-activated non-selective calcium permeant cation channel involved in detection of noxious chemical and thermal stimuli. Seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. May be involved in mediation of inflammatory pain and hyperalgesia. Sensitized by a phosphatidylinositol second messenger system activated by receptor tyrosine kinases, which involves PKC isozymes and PCL.
SIZE: 839 amino acids; 94938 Da
SUBUNIT: Self-associates. Probably homotetramer. May also form a heteromeric channel with TRPV3. Interacts with calmodulin, PRKCM and CSK. Interacts with PRKCG and NTRK1, probably by forming a trimeric complex (By similarity).
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein (By similarity).
TISSUE SPECIFICITY: Widely expressed at low levels. Expression is elevated in dorsal root ganglia. In skin, expressed in cutaneous sensory nerve fibers, mast cells, epidermal keratinocytes, dermal blood vessels, the inner root sheet and the infundibulum of hair follicles, differentiated sebocytes, sweat gland ducts, and the secretory portion of eccrine sweat glands (at protein level).DOMAIN:SwissProt: Q8NER1 The association domain (AD) is necessary for self- association (By similarity).
PTM: Phosphorylation by PKA reverses capsaicin-induced dephosphorylation at multiple sites, probably including Ser-117 as a major phosphorylation site. Phoshphorylation by CAMKII seems to regulate binding to vanilloids. Phosphorylated and modulated by PKCM and probably PKCZ. Dephosphorylation by calcineurin seems to lead to receptor desensitization and phosphorylation by CAMKII recovers activity (By similarity).
SIMILARITY: Belongs to the transient receptor family. TrpV subfamily. & Contains 3 ANK repeats.
MISCELLANEOUS: Responses evoked by low pH and heat, and capsaicin can be antagonized by capsazepine.

Application

Antibody blocking. Antiserum (Catalog No. AB5889) should be preincubated with peptide prior to incubation with tissue/sample. A 10-5 molar (M) peptide concentration is suggested to block antibody/antigen binding.

Optimal working dilution must be determined by the end user.

Physical form

Lyophilized in phosphate buffer, pH 7.4.

Storage and Stability

Store at -20°C or below in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service