Skip to Content
Merck
All Photos(1)

Key Documents

AG375

Sigma-Aldrich

µ Opioid Receptor control peptide

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€450.00

€450.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€450.00

About This Item

UNSPSC Code:
51111800
eCl@ss:
32160405
NACRES:
NA.41

€450.00


Estimated to ship on07 March 2025Details


Request a Bulk Order

concentration

2 mg/mL

Quality Level

technique(s)

inhibition assay: suitable

UniProt accession no.

Gene Information

General description

Mu opioid receptor control peptide for use with the anti-Mu Opioid Receptor Antibodies AB5509 and AB5511. Synthetic peptide corresponding to the C-terminus of rat mu Opioid receptor comprising residues 384-398.

Application

Antibody blocking studies (AB5511 and AB5509). Antibody should be pre-incubated with the peptide prior to incubation with the tissue/sample. It is suggested to pre-incubate for one hour at room temperature. A 10-6 to 10-5molar (M) peptide concentration is suggested to block antibody binding.

Optimal working dilutions must be determined by end user.

Physical form

Presented as a liquid.

Storage and Stability

Store at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service