Skip to Content
Merck
All Photos(1)

Key Documents

AG356

Sigma-Aldrich

P2X3, control peptide for AB5895 and AB5896

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€315.00

€315.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€315.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.77

€315.00


Estimated to ship on24 April 2025Details


Request a Bulk Order

biological source

rat

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

2 mg/mL

technique(s)

cell based assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

P2X3 control peptide for AB5895 and AB5986.

Application

Antibody blocking (AB5895 and AB5896)

Optimal working concentration must be determined by the end user.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service