Skip to Content
Merck
All Photos(2)

Key Documents

AG310

Sigma-Aldrich

Fluoro-Jade® B

Synonym(s):

FJB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Pricing and availability is not currently available.

form

powder

mol wt

Mw 681 Da

manufacturer/tradename

Fluoro-Jade®

technique(s)

immunofluorescence: suitable

fluorescence

λex 480 nm; λem 525 nm

General description

Fluoro-JadeⓇ B is a polyanionic fluorescein derivative which sensitively and specifically binds to degenerating neurons. It is a dark red power that has a green iridescence with excitation peak at 480 nm and emission peak at 525 nm. The filter used for visualizing Fluoro-JadeⓇ B is a fluorescein/FITC filter. Fluoro-JadeⓇ B can be used on most tissue section types and thicknesses including frozen, vibratomed cryostat or paraffin-embedded sections from 3-50 µm. Fluoro-JadeⓇ B is faster and more reliable than older methods (e.g. suppressed silver) for the unequivocal qualitative detection and quantitative measurement of both and fine scale neuronal degeneration.
N-terminal GST-tagged, recombinant human XIAP full length, expressed by baculovirus in Sf21 insect cells

Application

Appearance: Dark red powder with green iridescence.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Warning

Toxicity: Although the compound appears to be of low toxicity, it has not been extensively evaluated and therefore routine laboratory caution should be exercised. Not intended for human consumption.

Physical form

Filter System : Fluorescein / FITC
Solubility: Very soluble in water and bases, moderately soluble in alcohol and weak acids.

Storage and Stability

The powder should be stored well sealed at room temperature, preferable in a dessicator, due to its hygroscopic nature. The liquid stock solution (0.01%) in distilled water can be stored at 2-8°C for up to 3 months. The working solution (0.0004%) in 0.1% acetic acid should be prepared fresh and not be stored or reused.

Analysis Note

This layer chromatography using cellulose plates and a solvent system of n-propinol, water, and ammonium hydroxide (6:5:2) revealed the presence of two fluorescent isomers and two trace non-fluorescent bands. No amount of fluorescein or Fluoro-Jade was present

Legal Information

FLUORO-JADE is a registered trademark of Histo-Chem, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gislaine Olescowicz et al.
Neurotoxicity research, 38(2), 319-329 (2020-05-14)
Chronic treatment with agmatine, similarly to fluoxetine, may cause antidepressant-like effects mediated, at least in part, by the modulation of hippocampal plasticity. However, the ability of chronic treatment with agmatine to cause antidepressant-like effects associated with the modulation of mammalian
Jeffrey Landrigan et al.
Neurotoxicology, 76, 153-161 (2019-11-19)
Parkinson's disease (PD) is characterized by profound microglial driven inflammatory processes and the loss of dopamine neurons of the substantia nigra (SNc). Both microglia and dopamine neurons that are affected in the SNc are particularly vulnerable to environmental toxicants and
Maira Licia Foresti et al.
Epilepsy & behavior : E&B, 102, 106695-106695 (2019-12-01)
Neuroinflammation role on epileptogenesis has been the subject of increasing interest. Many studies showed elevation in cytokines and chemokines expression following seizures, such as, CCL2 protein (C-C motif ligand 2 chemokine) and its specific receptor, CCR2. In addition, recent studies
Naiara Silva Dourado et al.
Frontiers in aging neuroscience, 12, 119-119 (2020-06-06)
Neurodegenerative disorders (ND) are characterized by the progressive and irreversible loss of neurons. Alzheimer's Disease (AD) is the most incident age-related ND, in which the presence of a chronic inflammatory compound seems to be related to its pathogenesis. Different stimuli
Xiaolong Lin et al.
Frontiers in cellular neuroscience, 13, 567-567 (2020-01-11)
Previous reports showed that 2-(-2-benzofuranyl)-2-imidazoline (2-BFI) has antioxidant, anti-inflammatory and anti-apoptotic effects on neuroprotection in numerous disorders. However, the precise mechanisms remain elusive. The nuclear factor c factor 2 (Nrf2)/antioxidant response element (ARE) signaling pathway plays an important and essential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service