Skip to Content
Merck
All Photos(1)

Key Documents

AG201

Sigma-Aldrich

MCT4 Peptide

control for AB3550P, liquid

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

product name

MCT4, control peptide for AB3550P,

biological source

human

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

cell based assay: suitable

UniProt accession no.

shipped in

dry ice

Gene Information

human ... SLC16A3(9123)

General description

A 19 amino acid peptide sequence within the cytoplasmic domain of human MCT4 (1).

Application

For use in blocking the reactivity of AB3550P.

Optimal working dilution must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Ion Channels & Transporters

Physical form

Liquid in PBS containing 0.1% BSA and 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service