ACS5100
Amicon® Pro Purification System with 100kDa Amicon® Ultra-0.5 Device
Amicon Pro Purification System - A Centrifugal Tool collapsing protein purifcation with concentraion & buffer exchange.
Synonym(s):
100kDa Purification System, Amicon Pro System
About This Item
Recommended Products
material
Amicon Ultra styrene-butadiene device
Ultracel® regenerated cellulose membrane (Amicon Ultra-0.5; low binding)
polyethylene cap
polyethylene frit
polypropylene device (for exchange)
polypropylene frit
polypropylene tube (AU0.5; for collection)
polypropylene tube (for collection)
polypropylene tube (holder)
sterility
non-sterile
product line
Amicon®
manufacturer/tradename
Amicon® Pro
parameter
10 mL sample volume
technique(s)
buffer exchange: suitable
protein purification: suitable (antibody labeling)
protein purification: suitable (concentration)
protein purification: suitable (enrichment)
L
12 cm (4.7 in.)
diam.
2.97 cm
filtration area
1 cm2
working volume
10 mL
pore size
100 kDa NMWCO
shipped in
ambient
General description
Features and Benefits
- The Amicon® Pro Purification System eliminates the need for multiple sample transfers in the purification workflow thereby minimizing protein loss.
- Direct coupling to an Amicon Ultra-0.5 device provides simultaneous concentration during the elution phase, a feature particularly advantageous for depletion or enrichment applications
- The Amicon Pro Purification System offers highly efficient diafiltration, permitting buffer exchange and concentration of the final purified sample in a single 15-minute spin.
- Minimum Final Concentrate Volume: 15 L
Physical form
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Filtration methods and membrane filter selection in nanoparticle purification and production.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service