Skip to Content
Merck
All Photos(1)

Key Documents

ABT406

Sigma-Aldrich

Anti-LRRFIP1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Leucine-rich repeat flightless-interacting protein 1, LRR FLII-interacting protein 1, GC-binding factor 2, TAR RNA-interacting protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€370.00


Estimated to ship on11 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LRRFIP1(9208)

General description

Leucine-rich repeat flightless-interacting protein 1 (LRR FLII-interacting protein 1), also known as GC-binding factor 2 or TAR RNA-interacting protein, and encoded by the gene name LRRFIP1 or GCF2 or TRIP, is a transcriptional repressor that mainly plays a cytoskeletal role. LRRFIP1 is primarily localized in the cytoplasm and preferentially binds to GC-rich dsDNA, but will also bind directly to dsRNA as well. The RNA binding domain encompasses a lysine-rich motif. LRRFIP1 interacts with the mammalian Flightless I (Fli-I) and is a key component in the cytoskeletal regulation of platelet function. LRRFIP1 and the related protein LRRFIP2 may modulate canonical WNT signaling and mediate the IRF3-induced production of type I interferon via a β-catenindependent pathway.

Immunogen

KLH-conjugated synthetic peptide corresponding to human LRRFIP1.

Application

Anti-LRRFIP1 Antibody detects level of LRRFIP1 & has been published & validated for use in LRRFIP1.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Western Blotting in human colon tissue lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected LRRFIP1 in human colon tissue lysate.

Target description

~85 kDa observed. Uncharacterized bands may be observed in some lysate(s). Uniprot describes 4 isoforms produced by alternative splicing ranging between ~73 kDa and ~89 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service