Skip to Content
Merck
All Photos(1)

Key Documents

ABS543

Sigma-Aldrich

Anti-Colipase Antibody

serum, from rabbit

Synonym(s):

Colipase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€415.00

€415.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€415.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€415.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CLPS(1208)

General description

The protein Colipase is a co-enzyme needed for maximal enzyme activity of pancreatic lipase. It is a secreted protein produced by the pancreas that is originally secreted as an inactive proform, procolipase. Procolipase gets activated by cleavage by intestinal trypsin in the lumen of the small intestine. Once activated, Colipase′s function is to help overcome the inhibitory effect of bile salts on the lipase catalyzed hydrolysis of long-chain triglycerides that needs to occur within the duodenum. Colipase happens to have key hydrophobic amino acids that allow the complex of lipase and Colipase to adsorb to the lipid micelles, and this adsorption is essential for their digestion. Interestingly too, the procolipase propeptide also gives rise to the pentapeptide intestinal hormone Enterostatin, a small pentapeptide. Enterostatin acts to reduce food intake, in particular fat intake, when given to animals via injection or in to the brain.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human Colipase.

Application

Anti-Colipase Antibody | ABS543 is an antibody against Colipase Antibody for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected Colipase in purified protein (D′Silva, S., et al. (2007). Journal of Lipid Research. 48:2478-2484).
Western Blotting Analysis: A representative lot detected Colipase in mouse pancreas (Miller, R., et al. (2009). AM J Physiol Endocrinol Metab. 297:E856-E865).
Western Blotting Analysis: A representative lot detected Colipase in HEK293T cells transiently transfected to express Arg92 and Cys92 colipase (Xiao, X., et al. (2013). Journal of Lipid Research. 54:514-521).

Quality

Evaluated by Western Blotting in mouse pancreas tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Colipase in mouse pancreas tissue lysate.

Target description

~12 kDa observed

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service