Skip to Content
Merck
All Photos(2)

Key Documents

ABS435

Sigma-Aldrich

Anti-GNPDA2 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

GNPDA2, Glucosamine-6-phosphate deaminase 2, GNPI, Glucosamine-6-phosphate isomerase SB52, GNP2, GlcN6P deaminase 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€265.00

€265.00


Estimated to ship on10 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€265.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€265.00


Estimated to ship on10 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GNPDA2(132789)

General description

Glucosamine-6-phosphate isomerase 2 (GNPDA2), also known as Glucosamine-6-phosphate deaminase 2 and Glucosamine-6-phosphate isomerase SB52, is an allosteric enzyme that catalyzes the conversion of GNP to fructose-6-phosphate and ammonia. It consists of seven exons and six introns. GNPDA2 is ubiquitously expressed in most human tissues with high expression in testis, ovary, placenta, and heart.

Immunogen

KLH-conjugated linear peptide corresponding to human GNPDA2.

Application

Anti-GNPDA2 Antibody is a highly specific rabbit polyclonal antibody, that targets Glucosamine-6-phosphate isomerase 2 & has been tested in western blotting & ICC.
Immunocytochemistry Analysis: 4 µg/mL from a representative lot detected GNPDA2 in HEK293 cells.

Quality

Evaluated by Western Blotting in human kidney tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected GNPDA2 in 15 µg of human kidney tissue lysate.

Target description

~31 kDa observed. Uncharacterized bands may appear in some lysates. Uniprot describes 1 isoform at ~29 kDa and 2 isoforms at ~31 kDa

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service