Skip to Content
Merck
All Photos(3)

Key Documents

ABS2203

Sigma-Aldrich

Anti-UBE2A

from rabbit, purified by affinity chromatography

Synonym(s):

Ubiquitin-conjugating enzyme E2A, EC 2.3.2.23, E2 ubiquitin-conjugating enzyme A, RAD6 homolog A, HR6A, hHR6A, Ubiquitin carrier protein A, Ubiquitin-protein ligase A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€291.00

€291.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
€291.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€291.00


Estimated to ship on21 March 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... UBE2A(7319)

General description

Ubiquitin-conjugating enzyme E2 A (UniProt: P49459; also known as EC 2.3.2.23; E2 ubiquitin-conjugating enzyme A, RAD6 homolog A, HR6A, hHR6A, Ubiquitin carrier protein A, Ubiquitin-protein ligase A) is encoded by the UBE2A (also known as RAD6A) gene (GeneID: 7319) in human. UBE2A belongs to the ubiquitin-conjugating enzyme family and accepts ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. In association with the E3 enzyme BRE1, it plays a role in transcription regulation by catalyzing the mono-ubiquitination of histone H2B at Lys-120 that provides a specific tag for epigenetic transcriptional activation, elongation by RNA polymerase II, and telomeric silencing. It is also a prerequisite for H3K4me and H3K79me formation. Defects in UBE2A gene have been linked to mental retardation, X-linked, syndromic, Nascimento-type mental retardation that is characterized by significantly below average general intellectual functioning associated with impairments in adaptive behavior.

Specificity

This rabbit polyclonal antibody detects Ubiquitin-conjugating enzyme E2 A (UBE2A) in HeLa cells and Mouse Kidney.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to 14 amino acids from the C-terminal region of human Ubiquitin-conjugating enzyme E2 A (UBE2A).

Application

Anti-UBE2A Antibody, Cat. No. ABS2203, is a rabbit polyclonal antibody that detects UBE2A and has been tested for use Immunohistochemistry (Paraffin) and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:500 dilution from a representative lot detected UBE2A in mouse kidney tissue lysate.

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected UBE2A in human pancreas, human cervical cancer, and human uterus tissues.

Quality

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected UBE2A in HeLa cell lysate.

Target description

~17 kDa observed; 17.31 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wenjing Yan et al.
Nature communications, 14(1), 4798-4798 (2023-08-10)
UBA1 is the primary E1 ubiquitin-activating enzyme responsible for generation of activated ubiquitin required for ubiquitination, a process that regulates stability and function of numerous proteins. Decreased or insufficient ubiquitination can cause or drive aging and many diseases. Therefore, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service