Skip to Content
Merck
All Photos(2)

Key Documents

ABS2192

Sigma-Aldrich

Anti-mPTGES-1

from rabbit

Synonym(s):

Prostaglandin E synthase, EC:5.3.99.3, Microsomal glutathione S-transferase 1-like 1, MGST1-L1, Microsomal prostaglandin E synthase 1, MPGES-1, p53-induced gene 12 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€317.00

€317.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€317.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€317.00


Estimated to ship on30 April 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (based on high sequence homology)

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... PTGES(9536)

General description

Prostaglandin E synthase (UniProt: O14684; also known as EC:5.3.99.3, Microsomal glutathione S-transferase 1-like 1, MGST1-L1, Microsomal prostaglandin E synthase 1, MPGES-1, p53-induced gene 12 protein) is encoded by the PTGES gene (Gene ID: 9536) in human. mPGES -1 is a multi-pass membrane protein that is integral membrane, inducible and glutathione-dependent terminal synthase in the prostanoid biosynthesis pathway and acts downstream cyclooxygenases (COX) catalyzing the oxidoreduction of prostaglandin endoperoxide H2 (PGH2) to prostaglandin E2 (PGE2). Functionally, mPGES-1 is coupled to COX-2 and the induction of these two enzymes by pro-inflammatory cytokines, growth factors or endotoxin leads to a surge of PGE2 synthesis by inflammatory cells. It is strongly expressed with COX-2 in synovial fibroblasts and macrophages. In cases of osteoarthritis, mPGES-1 and COX-2 are strongly expressed in the chondrocytes and the synovial lining cells. mPGES-1 also play a role in the pathogenesis of ischemic stroke and many neurodegenerative diseases. In Alzheimer disease brains higher expression of both COX-2 and mPGES-1 is observed. Higher expression of mPGES has also been reported in several cancers and this higher expression has been correlated with vascular invasion and worse prognosis in colorectal cancer. A549 and DU145 xenografts lacking mPGES-1 display significantly reduced growth rates. (Ref.: Korotkova, M., and Jakobsson, PJ (2014). Basic Clin. Pharmacol. Toxicol. 114, 64-69).

Specificity

This polyclonal antibody specifically detects mPGES-1 in human.

Immunogen

His-tagged full-length recombinant human microsomal prostaglandin E synthase.

Application

Detect Prostaglandin E synthase using this rabbit polyclonal Anti-mPTGES-1, Cat. No. ABS2192 that has been tested in Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunofluorescence, and Western Blotting.
Immunohistochemistry Analysis: A 1:5,000 dilution from a representative lot detected mPTGES-1 in human tonsil and human placenta tissue.

Western Blotting Analysis: A representative lot detected mPTGES-1 in human chondrocytes (Tuure, L., et. al. (2015). Scand J Rheumatol. 44(1):74-9) and wild-type mixed glial cultures (Straccia, M., et. al. (2013). Glia. 61(10):1607-19).

Immunofluorescence Analysis: A representative lot detected mPTGES-1 in Wild-type mixed glial cultures (Tuure, L., et. al. (2015). Scand J Rheumatol. 44(1):74-9).

Western Blotting Analysis: A representative lot detected mPTGES-1 in PNE inhibitedmPGES-1proteinexpressioninIL-1 -stimulated SK-N-SH cells (Olajide, O.A., et. al. (2014). J Ethnopharmacol. 152(2):377-83).

Immunohistochemistry Analysis: A representative lot detected mPTGES-1 in fresh frozen brain tissue (Tuure, L., et. al. (2015). Scand J Rheumatol. 44(1):74-9).

Quality

Evaluated by Western Blotting in recombinant microsomal Prostaglandin E synthase 1 protein.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected mPTGES-1 in recombinant microsomal Prostaglandin E synthase 1 protein.

Target description

~17 kDa observed; 17.10 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit polyclonal antiserum without azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service