Skip to Content
Merck
All Photos(1)

Documents

ABS2087

Sigma-Aldrich

Anti-Mff

from rabbit

Synonym(s):

Mitochondrial fission factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... MFF(56947)

General description

Mitochondrial fission factor (UniProt: Q9GZY8; also known as Mff) is encoded by the MFF (also known as C2orf33, AD030, AD033, GL004) gene (Gene ID: 56947) in human. Mff is a single-pass type IV membrane protein that is highly expressed in heart, kidney, liver, brain, muscle, and stomach. Mff is a homodimeric protein, but may also interact with other proteins. It plays a role in mitochondrial and peroxisomal fission and promotes the recruitment and association of the fission mediator dynamin-related protein 1 (DNM1L) to the mitochondrial surface. It is also suggested to be involved in regulation of synaptic vesicle membrane dynamics by recruitment of DNM1L to clathrin-containing vesicles. Mff has a cytoplasmic domain (aa 1-322), a helical domain (aa 323-340), and a mitochondrial intermembrane domain (aa 341-342). Mff is shown to be an essential factor for mitochondrial recruitment of Drp1 during mitochondrial fission in mammalian cells and its down-regulation inhibits mitochondrial recruitment of Drp1 and induces mitochondria elongation. Also, exogenous expression of Mff is shown to induce extensive mitochondrial fission. Mutations in MFF gene can cause a form of encephalopathy that is characterized by delayed psychomotor development, severe hypotonia with inability to walk, and microcephaly. (Ref.: Otera, H., et al. (2010). J. Cell Biol. 191 (6); 1141 1158; Gandre-Babbe, S., and van der Bliek, AM. (2008). Mol. Biol. Cell. 19(6): 2402-2412).

Specificity

This rabbit polyclonal antibody detects mitochondrial fission factor (Mff) in human cells.

Immunogen

Epitope: unknown
His-taged full length human recombinant mitochondrial fission factor (Mff).

Application

Anti-Mff, Cat. No. ABS2087, is a rabbit polyclonal antibody that detects Mitochondrial fission factor (Mff) and has been tested for use in Immunocytochemistry, Immunofluorescence, and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A representative lot detected Mff in Western Blotting applications (Mishra, P., et. al. (2015). Cell Metab. 22(6):1033-44).

Immunofluorescence Analysis: A representative lot detected Mff in Immunofluorescence applications (Mishra, P., et. al. (2015). Cell Metab. 22(6):1033-44).

Quality

Evaluated by Immunocytochemistry in HeLa cells with Mitotracker (TM).

Immunocytochemistry Analysis: A 1:2,000 dilution of this antibody detected Mff in HeLa cells with Mitotracker (TM).

Target description

38.47 kDa calculated.

Physical form

Format: Unpurified
Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20Ā°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20Ā°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product? Ā 

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a productā€™s label following the words ā€˜Lotā€™ or ā€˜Batchā€™.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service