Skip to Content
Merck
All Photos(1)

Key Documents

ABS1633

Sigma-Aldrich

Anti-IST1 Homolog

serum, from rabbit

Synonym(s):

hIST1, Putative MAPK-activating protein PM28

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€280.00

€280.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€280.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

€280.00


Estimated to ship on12 March 2025Details


Request a Bulk Order

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology)

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... IST1(9798)

General description

IST1 homolog (UniProt: P53990; also known as hIST1, Putative MAPK-activating protein PM28) is encoded by the IST1 (also known as KIAA0174) gene (Gene ID: 9798) in human. IST1 is an ESCRT-III-like protein that is involved in specific functions of the ESCRT machinery. Is required for efficient abscission during cytokinesis. IST1 is also involved in recruiting VPS4A and/or VPS4B to the midbody of dividing cells. During late anaphase, it is involved in nuclear envelope reassembly and mitotic spindle disassembly together with the ESCRT-III complex: IST1 acts by mediating the recruitment of SPAST to the nuclear membrane, leading to microtubule severing. It is shown to regulate early endosomal tubulation together with the ESCRT-III complex by mediating the recruitment of SPAST. Six isoforms of IST1 homolog have been described that are generated by alternative splicing.

Specificity

This rabbit polyclonal antibody detects IST1 homolog in human cells. It targets an epitope within the first 189 amino acids from the N-terminal half.

Immunogen

A recombinant fragment corresponding to the first 189 amino acids from the N-terminal half of human IST1 homolog.

Application

Anti-IST1 Homolog, Cat. No. ABS1633,is a rabbit polyclonal antibody that detects IST1 homolog and has been tested for use in Immunofluorescence and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:5,000 dilution from a representative lot detected IST1 Homolog in 10 µg of HeLa cell lysate.
Western Blotting Analysis: A representative lot detected IST1 Homolog in Western Blotting applications (Bajorek, M., et. al. (2009). Nat Struct Mol Biol. 16(7):754-62; Bajorek, M., et. al. (2009). Mol Biol Cell. 20(5):1360-73).
Immunofluorescence Analysis: A representative lot detected IST1 Homolog in Immunofluorescence applications (Bajorek, M., et. al. (2009). Mol Biol Cell. 20(5):1360-73).

Quality

Evaluated by Western Blotting in Raji cell lysate.
Western Blotting Analysis: A 1:5,000 dilution of this antibody detected IST1 Homolog in 10 µg of Raji cell lysate.

Target description

~50 kDa observed; 39.75 kDa calculated.. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Rabbit polyclonal antiserum with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service