Skip to Content
Merck
All Photos(1)

Documents

ABS1013

Sigma-Aldrich

Anti-MTHFR Antibody

serum, from rabbit

Synonym(s):

Methylenetetrahydrofolate reductase, MTHFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse, porcine

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MTHFR(4524)

General description

Methylenetetrahydrofolate reductase or 5,10-methylenetetrahydrofolate reductase (EC 1.5.1.20; UniProt P42898) is encoded by the MTHFR gene (Gene ID 4524) in human. MTHFR catalyzes the reduction of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, the predominant circulatory form of folate and carbon donor for the re-methylation of homocysteine to methionine. Defects in MTHFR cause plasma homocysteine elevation or hperhomocysteinaemia, a risk factor for cerebrovascular, peripheral vascular, and coronary heart disease. Multiple transcription start sites and alternative splicing in the MTHFR gene result in two enzyme isoforms.

Specificity

Reacts with both isoforms (PMID 12370778).

Immunogen

Recombinant protein corresponding to human MTHFR.

Application

Detect MTHFR using this Anti-MTHFR Antibody validated for use in Western Blotting.
Research Category
Signaling
Research Sub Category
Signaling Neuroscience
Western Blotting Analysis: A representative lot detected MTHFR in mice plasma, kidney and brain (Chew, T.W., et al. (2011). J. Nutr. 141(8):1475-1481.).
Western Blotting Analysis: A representative lot detected MTHFR in porcine liver, human lymphocytes, human fetal kidney, human adult kidney, cells transfected with MTHFR (Rozen, R. U.S. Patent 6,833,243 B2, December 21, 2004).
Western Blotting Analysis: A representative lot detected MTHFR in HepG2, mouse liver, and RAW264.7 (Mikael, L.G., and Rozen, R. (2008). Cardiovasc. Res. 80(1):151-158).
Western Blotting Analysis: A representative lot detected recombinant MTHFR protein, as well as endogenous MTHFR in human kidney (both fetal & adult), fetal human lung, and adult human aorta (Tran, P., et al. (2002). Mamm. Genome. 13(9):483-492).

Quality

Evaluated by Western Blotting in RAW264.7 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected MTHFR in 10 µg of RAW264.7 cell lysate.

Target description

~70/77 kDa observed. Uncharacterized band(s) may appear in some lysates.

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service