Skip to Content
Merck
All Photos(1)

Key Documents

ABN993

Sigma-Aldrich

Anti-GHRH (Somatoliberin)

from sheep

Synonym(s):

Somatoliberin, Growth hormone-releasing factor, GRF, Growth hormone-releasing hormone

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€388.00

€388.00


Estimated to ship on19 March 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
€388.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€388.00


Estimated to ship on19 March 2025Details


Request a Bulk Order

biological source

sheep

Quality Level

antibody form

unpurified

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse

packaging

antibody small pack of 25 μL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
radioimmunoassay: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

rat ... Ghrh(29446)

General description

Somatoliberin (UniProt: P09916; also known as Growth hormone-releasing factor, GRF, Growth hormone-releasing hormone, GHRH) is encoded by the Ghrh gene (Gene ID: 29446) in rat. GHRH F is released by the hypothalamus and acts on the adenohypophyse to stimulate the secretion of growth hormone. The release of growth hormone from pituitary gland depends of the interplay between the stimulatory action of GHRH and inhibitory action of somatotropin release inhibiting factor (SRiF) or somatostatin. These two neurohormones are shown to be released in a reciprocal manner from the median eminence into the portal circulation and result in ultradian rhythmic release pattern of growth hormone.. Post-translational removal of the signal peptide (aa 1-19) and the two propeptide sequences (aa 20-30) and aa 75-104) releases the 43 aa mature Somatoliberin (aa 31-73), which functions as a ligand to stimulate growth hormone (GH) release from the pituitary. Variant receptors for GHRH have been found in several types of tumors, and antagonists of these receptors are shown to block the growth of the tumors. Defects in Ghrh gene are a known to cause of dwarfism, while hypersecretion of the encoded GHRH can lead to gigantism.

Specificity

This sheep polyclonal antibody detects somatolierin (GHRH) in mouse and rat brain sections. It targets an epitope within the first 29 amino acids.

Immunogen

BSA-conjugated synthetic linear peptide corresponding to the first 29 amino acids from rat Somatoliberin (GHRH).
Epitope: N-terminus

Application

Anti-GHRH (Somatoliberin) Antibody, Cat. No. ABN993, is a highly specific sheep polyclonal antibody that targets Somatoliberin (GHRH) and has been tested in Immunofluorescence, Immunohistochemistry (Paraffin), and Radioimmunoassay.
Radioimmunoassay Analysis: A representative lot detected GHRH (Somatoliberin) in Radioimmunoassay applications (Tannenbaum, G.S., et. al. (1990). Endocrinology. 127(6):3087-95).

Immunohistochemistry Analysis: A representative lot detected GHRH (Somatoliberin) in Immunohistochemistry applications (Tannenbaum, G.S., et. al. (1990). Endocrinology. 127(5):2551-60; Epelbaum, J., et. al. (1989). J Neuroendocrinol. 1(2):109-15).

Immunofluorescence Analysis: A representative lot detected GHRH (Somatoliberin) in Immunofluorescence applications (Stroh, T., et. al. (2009). J Neurosci. 29(25):8198-205).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry (Paraffin) in mouse brain lateral hypothalamic region.

Immunohistochemistry Analysis: A 1:2,000 dilution of this antibody detected GHRH (Somatoliberin) in mouse brain lateral hypothalamic region.

Target description

12.27 kDa calculated.

Physical form

Format: Unpurified
Sheep polyclonal antiserum with 0.01% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service